Vaccines for Children Program
The CDC’s Vaccines for Children program ensures that kids have a much better chance of getting their recommended vaccinations. The Vaccines available through the VFC Program are recommended by the Advisory Committee on Immunization and they help protect babies, young children, and adolescents from 16 diseases. To continue to help as many children as possible, the Vaccine Cold Chain is essential to keeping vaccines potent. Too much exposure to heat, cold, or light at any step in the cold chain can damage vaccines, resulting in loss of vaccine potency. Once lost, potency cannot be restored. One of the key elements for an effective cold chain is reliable temperature monitoring equipment.
Prevent Childhood Illness & Prevent Vaccine Losses
Think of your storage and monitoring equipment as an insurance policy to protect your patients from inadvertent administration of compromised vaccine, and your facility against costs of revaccination, replacement of expensive vaccines, and loss of patient confidence in your practice. For the best protection, your facility needs appropriate equipment that is set up correctly and maintained and repaired as needed.
"To fully ensure the safety of vaccines, the CDC recommends following equipment is recommended:
  • Stand-alone refrigerator(s) with enough space to accommodate your maximum inventory without crowding
  • Stand-alone freezer(s) with enough space to accommodate your maximum inventory without crowding
  • Digital data logger (DDL) with a current and valid Certificate of Calibration Testing (also known as a Report of Calibration) for each unit and at least one backup in case of a broken or malfunctioning device"
  Traceable® Wi-Fi Data Logging Refrigerator/Freezer Thermometers Compatible with TraceableLIVE® Cloud Service Traceable® Excursion-Trac™ Data Logging Refrigerator/Freezer Thermometers with Calibration Traceable® Memory-Loc™ Data Logging Thermometers with Calibration
  TraceableLIVE Excursion-Trac Memory-Loc
Datalogger
Meets CDC Recommendations
No Data Overwriting  
Cloud Notification    
Number of datapoints 6,000 onboard datapoints (cloud data memory is unlimited) 500,000 datapoints each
Traceable VFC-Compliant Products
VFC Traceable Products
Traceable products are calibrated to NIST standards to help you meet quality and regulatory recommendations. Most states recommend recalibrating your datalogger every 1-2 years. Is yours due? Request a quote today.
Digital Dataloggers (DDL)
Traceable Excursion-Trac™ Data Logging Thermometer with Calibration; 1 Bottle Probe
94460-07
Traceable Memory-Loc™ Datalogging Thermometer with Calibration; 1 Bottle Probe
94460-37
Traceable Wi-Fi Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE® Cloud Service; 2 Bottle Probes
99460-00
Traceable Excursion-Trac™ Data Logging Thermometer with Calibration; 2 Bottle Probes
94460-09
Traceable Memory-Loc™ Datalogging Thermometer with Calibration; 2 Bottle Probes
94460-39
Traceable Wi-Fi Data Logging Refrigerator/Freezer Thermometer Compatible with TraceableLIVE® Cloud Service; 1 Bottle and 1 Bullet Probe
99460-05
CDC Requirements
Traceable® Certificate supplied (Traceable to NIST)
Calibration Report Included
Annual Recalibration by ILAC/MRA accredited laboratory available
CDC Recommendations
Glycol encased buffered probe
Certificate of Calibration includes methods and procedure documentation
Certificate of Calibration includes stated uncertainties
Certificate of Calibration includes measurement results
Minimum/Maximum Memory
Capable of displaying current temperature and minimum/maximum memories
Minimum/Maximum Memory reset
Detachable probe
Adjustable High/Low Alarm (Audible and visual alarms)
Read storage unit temperatures without opening the door
±0.5°C (±1°F) Accuracy ±0.25°C ±0.25°C ±0.25°C
Low battery indicator
Temperature Excursion data
Data Logger with Continuous Monitoring and Active Display
Data Logger with user programmable logging intervals
Data Logger capable of storing 4,000 readings that will not overwrite when full ✔* ✔*
*Roughly 500,000 data points of memory (up to 300 years worth of data)
 ISO/IEC 17025 A2LA Accredited • ISO 17034 Certified Reference Material Producer 
Each individual state/local health department immunization program may have specific requirements—Consult your immunization program for more information.
Data represented above is accurate as of 03/7/2022. Recommendations and requirements are subject to change without prior notice.
Placement of a CDC.gov linking graphic or text link is to be used only as a marker to the CDC.gov home page. A link does not indicate any form of endorsement or approval from CDC, or the U.S. Department of Health and Human Services.