Sensidyne Gillian BDX II Abatement Air Sampler Kits
Convenient kits include everything you need for portable air sampling
  • Built-in rotameter and flow adjust offers flow rates of 500 to 3000 cc/min (0.50 to 3 LPM)
  • Compact, lightweight, and quiet sampling pump kits
  • Rechargeable battery pack system—ideal for portal use
  • Meets NIOSH 7400, 7082, 7105 and 7300
The BDXII Abatement Air Sampler is a dependable, low cost personal sampling pump. It is designed for use with asbestos and lead sampling. Meets NIOSH 7400, 7082, 7105 and 7300 specifications.
Choose between kits with one or five BDXII pumps with a Gilian or Zefon 5-station charger.
$330.00 - $2,416.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Battery Type Power (VAC) Description
Availability Pricing
COMPARE
5 rechargeable NiMH (included) 120 / 240 BDXII Air Sampling Pumps with Gilian 5 Station Charger; 120 to 240V
$2,416.00USD / Pkg of 5
COMPARE
5 rechargeable NiMH (included) 120 / 240 BDXII Air Sampling Pumps with 5 Station Charger; 120 to 240V
$1,843.00USD / Each
COMPARE
1 Rechargeable NiMH (included) 120 BDXII Air Sampling Pump Starter Kit; 120 VAC
$330.00USD / Each