Zahn Viscosity Cups
Stainless steel cup with precsion-drilled orifice for quick, easy use in shops, factories, and laboratories.
$261.00 - $316.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Cup Number
Availability Pricing
BYK-Gardner PV-8202 Zahn Viscosity Cup, number 2, 30 to 230 cSt, 22 to 80 sec EW-08711-32 Mfr # PV-8202
COMPARE
2
$316.00USD / Each
BYK-Gardner PV-8204 Zahn Viscosity Cup, number 4, 230 to 1100 cSt, 20 to 80 sec EW-08711-36 Mfr # PV-8204
COMPARE
4
$261.00USD / Each