YSI Pro20i Dissolved Oxygen Meters
Automatic barometric pressure compensation ensures accurate measurements
  • Get DO calibrations within 3 seconds with a touch of a button
  • Store up to 50 data sets—no need to write down data
  • Easily read measurements in any light with backlit graphic display and keypad
  • Rubber overmolded case provides waterproof IP67-rated protection
  • Prestretched membrane caps make installing easy
The rugged, easy-to-use portable dissolved oxygen (DO) meters can be quickly calibrated with a push of a button for fast, accurate measurements and response times. The glow-in-the-dark keypad allows for easy navigation in low-light areas, while the display is viewable in all lighting conditions, even direct sunlight while wearing polarized sunglasses.
The meters are available with either a classic polarographic or “instant-on” galvanic user-replaceable DO probe with screw-on membrane caps for ease of maintenance. The cap membrane is prestretched at the factory making it easier to install. The membrane material requires less stirring and offers faster response time.
The rubber over-molded case provides a sturdy hand grip and protects meters while in the field. Meters have a IP67 waterproof rating even without the battery lid. Additional features include adjustable right- and left-handed strap, battery-life indicator, and multiple language display (English, Spanish, French, and German).
$1,092.00 - $1,185.50USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI Pro20i Dissolved Oxygen Meter, 1-m Cable, Polarographic EW-59353-40 Mfr # 607130
COMPARE
Dissolved Oxygen Meter, 1-m Cable, Polarographic
$1,092.00USD / Each
YSI Pro20i Dissolved Oxygen Meter, 1-m Cable, Galvanic EW-59353-41 Mfr # 607131
COMPARE
Dissolved Oxygen Meter, 1-m Cable, Galvanic
$1,092.00USD / Each
YSI Pro20i Dissolved Oxygen Meter, 4-m Cable, Polarographic EW-59353-42 Mfr # 607125
COMPARE
Dissolved Oxygen Meter, 4-m Cable, Polarographic
$1,118.00USD / Each
YSI Pro20i Dissolved Oxygen Meter, 4-m Cable, Galvanic EW-59353-43 Mfr # 607129
COMPARE
Dissolved Oxygen Meter, 4-m Cable, Galvanic
$1,118.00USD / Each
YSI Pro20i Dissolved Oxygen Meter, 10-m Cable, Polarographic EW-59353-44 Mfr # 607132
COMPARE
Dissolved Oxygen Meter, 10-m Cable, Polarographic
$1,185.50USD / Each
YSI Pro20i Dissolved Oxygen Meter, 10-m Cable, Galvanic EW-59353-45 Mfr # 607133
COMPARE
Dissolved Oxygen Meter, 10-m Cable, Galvanic
$1,185.50USD / Each