YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assemblies with Conductivity Sensor
Get lab-quality results at the source
  • Advanced high-accuracy optical dissolved oxygen probe requires little maintenance Smart probes store calibration information for up to one year saving you time and money
Interchangeable digital "smart" probes store calibration data for a year, saving you time and calibration solution costs—no need to recalibrate if connected to different meters. Convenient cable management kits are included with probes featuring cable lengths from 13-ft (4-m) to 100-ft (30-m)
$1,489.25 - $1,724.25USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 1 m Cable EW-59353-78 Mfr # 627150-1
COMPARE
Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 1 m Cable
$1,489.25USD / Each
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 4 m Cable EW-59353-79 Mfr # 627150-4
COMPARE
Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 4 m Cable
$1,489.25USD / Each
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 10 m Cable EW-59353-80 Mfr # 627150-10
COMPARE
Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 10 m Cable
$1,578.00USD / Each
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 20 m Cable EW-59353-81 Mfr # 627150-20
COMPARE
Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 20 m Cable
$1,609.50USD / Each
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 30 m Cable EW-59353-82 Mfr # 627150-30
COMPARE
Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 30 m Cable
$1,640.75USD / Each
YSI ODO/CT Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 50 m Cable EW-59353-83 Mfr # 627150-50
COMPARE
Optical Dissolved Oxygen Probe and Cable Assembly with Conductivity Sensor, 50 m Cable
$1,724.25USD / Each