YSI MultiLab DO/BOD Benchtop Meter Kits
Meets US EPA-approved methods for dissolved oxygen and BOD
  • Large, easy-to-read graphical display with backlight
  • Digital plug-and-play sensors for optical DO, BOD, and pH
  • CMC function monitors sensors measuring range against calibration range
  • Data logging with GLP time-and-date tracebility and USB connectivity
  • BOD Analyst Pro software automatically calculates BOD values
YSI Multilab dissolved oxygen and pH/dissoved oxygen meters help you comply with US EPA regulations. These low-profile lab meters are ideal for highly accurate measuring of DO, BOD, and pH.
The IP64-rated waterproof meters feature intuitive menu-driven interface, large graphical display with backlight, manual and automatic data logging with GLP tracability, and convenient USB data transfer.
Meter uses Intelligent Design Sensors (IDS) that digitally store their serial number, unique ID, and calibration data. The sensors can be moved from one meter to another and transfer their calibration data.
$2,055.00 - $2,688.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Min Dissolved Oxygen Description
Availability Pricing
YSI MultiLab BOD Benchtop Lab Kit, with optical DO/BOD probe EW-05510-02 Mfr # 626570
COMPARE
40 0 to 50 ppm; 0 to 500% saturation BOD Benchtop Lab Kit, with optical DO/BOD probe
$2,055.00USD / Each
YSI 626571 MultiLab Advanced BOD Benchtop Lab Kit EW-05510-04 Mfr # 626571
COMPARE
40 0 to 50 ppm; 0 to 500% saturation MultiLab Advanced BOD Benchtop Lab Kit
$2,688.00USD / Each
YSI 626573 MultiLab BOD/pH Benchtop Lab Kit, w/ optical DO/BOD probe and pH electrode EW-05510-06 Mfr # 626573
COMPARE
40 0 to 50 ppm; 0 to 500% saturation MultiLab BOD/pH Benchtop Lab Kit, w/ optical DO/BOD probe and pH electrode
$2,652.00USD / Each