YSI EcoSense DO200A Dissolved Oxygen Meter
Accurate, economical DO measurements
  • Automatic temperature compensation
  • Manual input for salinity and barometric pressure compensation
  • Easy-to-replace screw-on cap membranes
The DO200A combines low cost with a rugged, waterproof design--making it ideal for spot testing in wastewater, surface water, or aquaculture. Meter features easy-to-use interface, one-hand operation, and results in mg/L (ppm) or % saturation with temperature, salinity and barometric pressure compensation. Meter also has easy-to-replace membrane cap, auto shutoff, low battery indicator, and nonvolatile 50 data sets memory with calibration.
$381.25 - $824.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
YSI DO200 Kit EcoSense DO 200 Dissolved Oxygen Meter Only EW-53014-30 Mfr # 606077
COMPARE
EcoSense DO 200 Dissolved Oxygen Meter Only
$381.25USD / Each
YSI DO200 Kit EcoSense DO 200 Dissolved Oxygen Meter Kit
Always in Stock
EW-53014-32 Mfr # 606072
COMPARE
EcoSense DO 200 Dissolved Oxygen Meter Kit
$824.00USD / Each