WTW Conductivity Probes
Four-electrode conductivity cells offer a high degree of accuracy over wide measuring range
  • Do not need platinization
WTW conductivity probes to suit your conductivity measurement needs.
$637.76 - $872.14USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
WTW 301961 LR 325/01 conductivity probe (Ultra-pure water) EW-19706-62 Mfr # 301961
COMPARE
TetraCon 4-electrode conductivity probe, 3-m cable, waterproof DIN connector
$742.61USD / Each
WTW 301970 TetraCon 4-electrode conductivity probe, 3-m cable, waterproof DIN connector EW-19707-80 Mfr # 301970
COMPARE
TetraCon 4-electrode conductivity probe, 3-m cable, waterproof DIN connector
$637.76USD / Each
WTW 301972 TetraCon 4-electrode conductivity probe, 10-m cable, waterproof DIN connector EW-19707-82 Mfr # 301972
COMPARE
4-electrode conductivity probe, 10-m cable, waterproof DIN connector
$727.81USD / Each
WTW 301974 TetraCon 4-electrode conductivity probe, 20-m cable, waterproof DIN connector EW-19707-84 Mfr # 301974
COMPARE
$872.14USD / Each