This product is now discontinued, View Related Products
WIREMOLD / LEGRAND Hard-Wired Surge Protectors
Provide up to 160,000 A surge protection per phase
  • NEMA 4X for indoor and outdoor installations
These surge devices protect sensitive electronic equipment from external or internal transient activity. The water-tight, fire- and corrosion-resistant plastic housing provides protection for indoor and outdoor mounting. Easily drill or punch the enclosure for conduit or cable access installation. Units provide excellent protection when installed at main service panels, sub panels or at specific pieces of equipment. All modes (L-N, L-L, N-G, and L-G) feature 40,000 A (26873-00 to –06) or 80,000 A (26873-10 to –16) surge capacity. A green LED indicates operating protection present per phase. Optional flush-mount cover plate (sold separately below) provides a finished look to installation on sheetrock or a firewall.
Call For Price
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Max Current Capacity (A/phase) Surge Suppression
Availability Pricing
Wiremold/Legrand PA120T Single Phase, 3 Wire Surge Protector (80,000 A per phase), 120/240 VAC EW-26873-00 Mfr # PA120T
COMPARE
80000 80,000 A per phase
Call For Price
Wiremold/Legrand PA120Y Three Phase, 4 Wire Surge Protector (80,000 A per phase), 120/208 VAC EW-26873-02 Mfr # PA120Y
COMPARE
80000 80,000 A per phase
Call For Price
Wiremold/Legrand PA277Y Three Phase, 4 Wire Surge Protector (80,000 A per phase), 277/480 VAC EW-26873-04 Mfr # PA277Y
COMPARE
80000 80,000 A per phase
Call For Price
Wiremold/Legrand PA240DCT Hi-Leg Delta Surge Protector (80,000 A per phase), 120/240/120 VAC EW-26873-06 Mfr # PA240DCT
COMPARE
80000 80,000 A per phase
Call For Price
Wiremold/Legrand PB120T Single Phase, 3 Wire Surge Protector (160,000 A per phase), 120/240 VAC EW-26873-10 Mfr # PB120T
COMPARE
160000 160,000 A per phase
Call For Price
Wiremold/Legrand PB120Y Three Phase, 4 Wire Surge Protector (160,000 A per phase), 120/208 VAC EW-26873-12 Mfr # PB120Y
COMPARE
160000 160,000 A per phase
Call For Price
Wiremold/Legrand PB277Y Three Phase, 4 Wire Surge Protector (160,000 A per phase), 277/480 VAC EW-26873-14 Mfr # PB277Y
COMPARE
160000 160,000 A per phase
Call For Price
Wiremold/Legrand PB240DCT Hi-Leg Delta Surge Protector (160,000 A per phase), 120/240/120 VAC EW-26873-16 Mfr # PB240DCT
COMPARE
160000 160,000 A per phase
Call For Price
Showing 1- 8 of 8