Wika Standard Pressure Gauges 111.12
Our Standard Pressure Gauges are designed for long and reliable service under rugged conditions. Some typical applications are for pumps, hydraulic and pneumatic systems, compressors, and many more applications where liquid and gases do not corrode brass. The accuracy of the gauges is ± 3/2/3% of span in all sizes and they are available from 30 psi to 5000 psi. Vacuum ranges and compound ranges are available upon request.
$22.20 - $34.40USD / Each
13 variations of this product are available.
See All Product Options Below
13 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Accuracy Dial Size (in) Max Pressure (PSI)
Availability Pricing
Wika 9690463 1.5" Pressure Gauge, 0 to 100 psi, Back Mount EW-68430-12 Mfr # 9690463
COMPARE
±3-2-3% full-scale 2 100
$22.21USD / Each
Wika 9690714 2" Pressure Gauge, 0 to 200 psi, Back Mount EW-68430-33 Mfr # 9690714
COMPARE
±3-2-3% full-scale 2 200
$22.20USD / Each
Wika 111.12 1.5 1.5" Standard Pressure Gauge, 0 to 60 psi, Back Mount EW-68433-14 Mfr # 9690225
COMPARE
±3-2-3% full-scale 1.5 60
$23.93USD / Each
Wika 111.12 1.5 1.5" Standard Pressure Gauge, 0 to 100 psi, Back Mount EW-68433-15 Mfr # 9690234
COMPARE
±3-2-3% full-scale 1.5 100
$23.93USD / Each
Wika 111.12 1.5 1.5" Standard Pressure Gauge, 0 to 200 psi, Back Mount EW-68433-16 Mfr # 9690250
COMPARE
±3-2-3% full-scale 1.5 200
$23.93USD / Each
Wika 111.12 2 2" Standard Pressure Gauge, 0 to 100 psi, Back Mount EW-68433-18 Mfr # 9690692
COMPARE
±3-2-3% full-scale 2 100
$22.20USD / Each
Wika 111.12 1.5 1.5" Standard Pressure Gauge, 0 to 30 psi, Back Mount EW-68433-25 Mfr # 9690217
COMPARE
±3-2-3% full-scale 1.5 30
$23.93USD / Each
Wika 111.12 1.5 1.5" Standard Pressure Gauge, 0 to 160 psi, Back Mount EW-68433-26 Mfr # 9690242
COMPARE
±3-2-3% full-scale 1.5 160
$23.93USD / Each
Wika 111.12 2 2" Standard Pressure Gauge, 0 to 160 psi, Back Mount; 1/4"NPT EW-68433-28 Mfr # 9690705
COMPARE
±3-2-3% full-scale 2 160
$22.20USD / Each
Wika 111.12 2 2" Standard Pressure Gauge, 0 to 160 psi, Back Mount; 1/8"NPT EW-68434-14 Mfr # 9690471
COMPARE
±3-2-3% full-scale 2 160
$22.21USD / Each
Wika 111.12 2 2" Standard Vacuum Gauge, 30 to 0" Hg, Back Mount EW-68434-15 Mfr # 9690586
COMPARE
±3-2-3% full-scale 2 14.7
$27.00USD / Each
Wika 111.12 2.5 2.5" Standard Vacuum Gauge, 30 to 0" Hg, Back Mount EW-68434-17 Mfr # 52813116
COMPARE
±3-2-3% full-scale 2.5 14.7
$34.40USD / Each
Wika 111.12 2.5 2.5" Standard Pressure Gauge, 0 to 100 psi, Back Mount EW-68434-19 Mfr # 9691141
COMPARE
±3-2-3% full-scale 2.5 100
$24.69USD / Each
Showing 1- 13 of 13