Whirl-Pak Sampling Bags with White Labels
Keep samples safe—easy-seal bags hold solids and liquids
  • Quick and easy sealing—flip bag around wire/tape and bend to seal
  • No more poking—puncture-proof tabs keep wire intact even after extensive use
All bags are sterilized after manufacturing using ethylene oxide gas at an independent ISO-certified facility. Exceptionally clear film and superior strength are achieved by using co-extrusion low-density and linear low-density, virgin polyethylene.
The white tape closure makes sample identification easier and more convenient. Capacity is measured in fluid ounces when the bag is closed, and tab is folded over three times. Volume and dimensions are approximate, bags should not be used at temperatures above 180° F (82° C). Bags can be frozen to any temperature, even liquid nitrogen.
$110.50 - $382.50USD / Box of 250
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (oz) Thickness (mil) Description
Availability Pricing
Whirl-Pak B01064WA Sterile Sampling Bag with White Labeling Area, 2 oz; 500/Bx EW-06499-60 Mfr # B01064
COMPARE
2 2.5 Sterile sampling bag with white labeling area, 2 oz
$110.50USD / Box of 500
Whirl-Pak B01062WA Sterile Sampling Bag with White Labeling Area, 4 oz; 500/Bx
Always in Stock
EW-06499-70 Mfr # B01062
COMPARE
4 2.25 Sterile sampling bag with white labeling area, 4 oz
$113.75USD / Box of 500
Whirl-Pak B01489WA Sterile Sampling Bag with White Labeling Area, 7oz; 500/Bx EW-06499-72 Mfr # B01489
COMPARE
7 2.5 Sterile Sampling Bags with White Labeling Area, 7oz 500/BX
$142.25USD / Box of 500
Whirl-Pak B01490WA Sterile Sampling Bag with White Labeling Area, 13oz: 500/Bx EW-06499-74 Mfr # B01490
COMPARE
13 2.5 Sterile Sampling Bags with White Label Area, 13oz, 500/bx
$186.75USD / Box of 500
Whirl-Pak B01065WA Sterile Sampling Bag with White Labeling Area, 18 oz; 500/Bx
Always in Stock
EW-06499-80 Mfr # B01065
COMPARE
18 2.5 Sterile sampling bag with white labeling area, 18 oz
$160.00USD / Box of 500
Whirl-Pak B01297WA Sterile Sampling Bag with White Labeling Area, 24 oz; 500/Bx EW-06499-85 Mfr # B01297
COMPARE
24 3 Sterile sampling bag with white labeling area, 24 oz
$210.00USD / Box of 500
Whirl-Pak B01491WA Sterile Sampling Bag with White Labeling Area, 27oz; 500/Bx EW-06499-87 Mfr # B01491
COMPARE
27 3 Sterile Sampling Bags with White Label Area, 27oz 500/bx
$244.00USD / Box of 500
Whirl-Pak B01515WA Sterile Sampling Bag with White Labeling Area, 69oz; 500/Bx EW-06499-89 Mfr # B01515
COMPARE
69 3 Sterile Sampling Bags with White Labeling Area, 69oz 500/bx
$382.50USD / Box of 500
Whirl-Pak B01445WA Sterile Sampling Bag with White Labeling Area, 92 oz; 250/Bx EW-06499-94 Mfr # B01445
COMPARE
92 4 Sterile Sampling Bags with White Label Area, 92 oz 250/bx
$312.50USD / Box of 250
Whirl-Pak B01446WA Sterile Sampling Bag with White Labeling Area, 123oz; 250/Bx EW-06499-98 Mfr # B01446
COMPARE
123 4 Sterile Sampling Bags with White Label Area, 123oz 250/bx
$373.75USD / Box of 250
Whirl-Pak B01447WA Sterile Sampling Bag with White Labeling Area, 184 oz; 100/Bx EW-06499-99 Mfr # B01447
COMPARE
184 4 Sterile Sampling Bags with White Label Area, 184 oz, 100/bx
$237.75USD / Box of 100
Showing 1- 11 of 11