Whatman™ Polycap™ TC Filter Capsules, PES
Pre-sterilized polyethersulfone (PES) membrane
  • Select capsules with filling bell for cleanliness assurance during filling
These capsules contain two layers of polyethersulfone (PES) membrane, which is inherently hydrophilic, low in extractables, and offers excellent flow rates. The membrane also exhibits low protein binding characteristics making it ideal for use with biologically sensitive solutions. The device is supplied presterilized and has a vent to evacuate any air to allow fast filtration. The outlet of the capsule is protected to prevent touch contamination during setup and has standard hose barb connections.
$110.50 - $334.00USD / Pkg of 1
11 variations of this product are available.
See All Product Options Below
11 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Whatman 6714-3604 Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 550 cm2 EW-29701-46 Mfr # 6714-3604
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 550 cm2
$117.00USD / Pkg of 1
Whatman 6715-3602 Polycap TC Filter Capsule, PES, Sterile, 0.2um, 550 cm2, Filling Bell EW-29701-48 Mfr # 6715-3602
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.2um, 550 cm2, Filling Bell
$140.50USD / Pkg of 1
Whatman 6715-3682 Polycap TC Filter Capsule, PES, Sterile, 0.8/0.2um, 380 cm2, Filling Bell EW-29701-50 Mfr # 6715-3682
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.8/0.2um, 380 cm2, Filling Bell
$110.50USD / Pkg of 1
Whatman 6715-3604 Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 550 cm2, Filling Bell EW-29701-52 Mfr # 6715-3604
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 550 cm2, Filling Bell
$124.50USD / Pkg of 1
Whatman 6714-7502 Polycap TC Filter Capsule, PES, Sterile, 0.2um, 1100 cm2 EW-29701-54 Mfr # 6714-7502
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.2um, 1100 cm2
$207.50USD / Pkg of 1
Whatman 6715-7502 Polycap TC Filter Capsule, PES, Sterile, 0.2um, 1100 cm2, Filling Bell EW-29701-58 Mfr # 6715-7502
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.2um, 1100 cm2, Filling Bell
$155.75USD / Pkg of 1
Whatman 6715-7582 Polycap TC Filter Capsule, PES, Sterile, 0.8/0.2um, 760 cm2, Filling Bell EW-29701-60 Mfr # 6715-7582
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.8/0.2um, 760 cm2, Filling Bell
$169.50USD / Pkg of 1
Whatman 6717-9502 Polycap TC Filter Capsule, PES, Sterile, 0.2um, 2200 cm EW-29701-64 Mfr # 6717-9502
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.2um, 2200 cm
$259.25USD / Pkg of 1
Whatman 6717-9504 Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 2200 cm EW-29701-66 Mfr # 6717-9504
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.45/0.65um, 2200 cm
$235.50USD / Pkg of 1
Whatman 6718-9502 Polycap TC Filter Capsule, PES, Sterile, 0.2um, 2200 cm, Filling Bell EW-29701-68 Mfr # 6718-9502
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.2um, 2200 cm, Filling Bell
$334.00USD / Pkg of 1
Whatman 6718-9582 Polycap TC Filter Capsule, PES, Sterile, 0.8/0.2um, 1140 cm2, Filling Bell EW-29701-70 Mfr # 6718-9582
COMPARE
Polycap TC Filter Capsule, PES, Sterile, 0.8/0.2um, 1140 cm2, Filling Bell
$194.00USD / Pkg of 1
Showing 1- 11 of 11