Wearwell PermaTack Tacky Mats

Prevent debris and bacteria from entering sterile areas
  • Reusable and economical — replace your multilayer adhesive mats
  • Permanently tacky surface removes particles off shoes and wheels; contaminates remain trapped until mat is cleaned
  • Removes 98 to 100% of particles if each foot touches the mat three times
  • Regenerate the tack by simply cleaning the mat and allowing it to dry
  • Adhesive backing allows easy installation and prevents mat movement
  • Surface is static dissipative; RTT = 1 x 108 to 1 x 1010 ohms
$683.80 - $3,057.80USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Length (ft) Length (in) Width (in)
Availability Pricing
Wearwell PermaTack Tacky Mat, 27 in x 48 in EW-84580-20 Mfr # 092.18x27x48BL
COMPARE
4 48 27
$683.80USD / Each
Wearwell PermaTack Tacky Mat, 27" x 72", Blue EW-84580-21 Mfr # 092.18x27x72BL
COMPARE
6 72 27
$973.80USD / Each
Wearwell PermaTack Tacky Mat, 36" x 54", Blue EW-84580-22 Mfr # 092.18x36x54BL
COMPARE
4.5 54 36
$985.80USD / Each
Wearwell PermaTack Tacky Mat, 27" x 10 ft EW-84580-23 Mfr # 092.18x27x10BL
COMPARE
10 120 27
$1,577.80USD / Each
Wearwell PermaTack Tacky Mat, 54" x 10 ft EW-84580-24 Mfr # 092.18x54x10BL
COMPARE
10 120 54
$3,057.80USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat