Vortex-Genie 1 Shakers
Enhanced vortex action for maximum high-speed touch mixing
  • Solid metal casing with rubber feet prevents shaker from ''walking'' across your benchtop
Vortex - Genie® 1 Shaker has a single high-speed mixing action that eliminates operator error for highly reproducible results.
$441.25 - $447.25USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Power (VAC)
Availability Pricing
Scientific Industries Vortex-Genie SI-0136 1 Shaker, 600 to 3200 rpm, 120 VAC EW-04724-10 Mfr # SI-0136
COMPARE
120
$441.25USD / Each
Scientific Industries Vortex-Genie SI-0146 1 Shaker, 600 to 2700 rpm, 230 VAC EW-04724-15 Mfr # SI-0146
COMPARE
240
$447.25USD / Each