Vistek Balance Isolation Platforms
Reduce the transmission of vibrations to balances and other sensitive equipment
  • Portable and lightweight–locate your balance on any countertop or table
  • Maintains balance accuracy even in the presence of operating equipment
  • Balances to hold calibration longer and load cells are protected from damage
Lightweight and easy to set up, the platform isolates balances and other sensitive instruments and equipment from vibrations and mechanical shock—ideal for balances, petri dishes, and shakers.
Sturdy, gray PVC platform features a non-stick neoprene surface supported by three or four vibration isolation bearings. These advanced isolation bearings protect balances and other sensitive equipment from ambient vibrations and mechanical shocks.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Platform Length (in) Platform Width (in) Description
Availability Pricing
Newport LIP275-0810-01 Balance Isolation Platform, 8" W x 10" D EW-11500-01 Mfr # LIP275-0810-01
COMPARE
15 10 Balance Isolation Platform, 8\" W x 10\" D
Call For Price
Newport LIP275-1015-01 Balance Isolation Platform, 10" W x 15" D EW-11500-02 Mfr # LIP275-1015-01
COMPARE
15 10 Balance Isolation Platform, 10\" W x 15\" D