Vibration Meters with Electronic Stethoscope
Versatile meter for vibration analysis
  • Ideal for troubleshooting and preventive maintenance programs — use on motors, gearboxes, pumps, compressors, and fans
Use the vibration meter and electronic stethescope in combination with the accelerometer vibration sensor and headphones to quickly locate noise and chatter in your machinery. Vibration severity chart on front of meter provides instant status by relating the digital reading to machinery applications per ISO 10816-1. Software lets you store, compare, and graph your measurements.
Software System Requirements: System: 486 computer or better, CD-ROM, running Windows® 95/98, XP and NT 4.0; Memory: 8 MB RAM; Hard disk space: 100 MB minimum
$1,208.00 - $1,442.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Monarch 6400-012 Vibration Meter with Electronic Stethoscope EW-98834-00 Mfr # 6400-012
COMPARE
Vibration Meter with Electronic Stethoscope
$1,208.00USD / Each
Monarch 6400-011 Vibration Meter with Sensor Pk EW-98834-03 Mfr # 6400-011
COMPARE
Vibration Meter with Sensor Pk
$1,442.00USD / Each