Vector Laboratories EndTag™ DNA Labeling Systems
More efficient labeling of short oligonucleotides
  • Simple—get reliable, uniform labeling in approximately one hour
  • Diverse choice of labels: fluorophores, biotin, haptens, and affinity ligands (not included)
EndTag DNA labeling systems are ideal for applications where an internal label might interfere with hybridization or sequence specific protein binding. Systems make labeling of short oligonucleotides more efficient and is an economical alternative to having labels inserted during synthesis. Both EndTag systems enable the covalent attachment of a variety of fluorescent dyes, haptens, or affinity tags to the respective ends of the nucleic acids using thiol-specific chemistry. Labels containing thiol-reactive groups can easily be incorporated. Note: Reactions require labeling reagents (Biotin maleimide, DNP maleimide, Fluorescein maleimide, Fucose maleimide, Texas Red® maleimide).
$215.00 - $314.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Vector Laboratories EndTag™ 5' Nucleic Acid Labeling Kit EW-93952-13 Mfr # MB-9001
COMPARE
5' Nucleic Acid Labeling Kit
$215.00USD / Each
Vector Laboratories EndTag™ 3' Nucleic Acid Labeling Kit EW-93952-14 Mfr # MB-9002
COMPARE
3' Nucleic Acid Labeling Kit
$314.00USD / Each