Vector Laboratories Alkaline Phosphatase Substrate Kits

Successfully label multiple antigens in same tissue section
  • Dropper bottles simplify handling of chromogens
  • No wait times for mixing and dissolving powders or tablets
These enzyme substrate kits for use with alkaline phosphatase detection systems offer a wide variety of applications. Numerous color choices are available for staining antigens in pigmented tissues and melanomas, as well as for localizing multiple antigens in the same tissue section. Viewing options for the substrates include brightfield, fluorescence, darkfield, electron microscopy, and spectral imaging.
$211.50 - $245.00USD / Each

5 variations of this product are available.

See All Product Options Below
5 PRODUCT OPTIONS
Compareitem specifications
Color Volume (mL) Description
Availability Pricing
Vector Laboratories SK-5100 Red Substrate Kit, Magenta, 200 ml EW-93952-02 Mfr # SK-5100
COMPARE
Magenta 200 Red Substrate Kit, Magenta, 200 ml
$244.00USD / Each
Vector Laboratories ImmPACT Vector Red Substrate Kit, Red, 120 ml EW-93952-03 Mfr # SK-5105
COMPARE
Red 120 Vector Red Substrate Kit, Red, 120 ml
$211.50USD / Each
Vector Laboratories SK-5200 Black Substrate Kit, Brown-Black, 200 ml EW-93952-04 Mfr # SK-5200
COMPARE
Brown-black 200 Black Substrate Kit, Brown-black, 200 ml
$229.00USD / Each
Vector Laboratories SK-5300 Blue Substrate Kit, Blue, 200 ml EW-93952-05 Mfr # SK-5300
COMPARE
Blue 200 Blue Substrate Kit, Blue, 200 ml
$245.00USD / Each
Vector Laboratories SK-5400 BCIP/NBT Substrate Kit, Indigo, 200 ml EW-93952-06 Mfr # SK-5400
COMPARE
Indigo 200 BCIP/NBT Substrate Kit, Indigo, 200 ml
$229.00USD / Each
Showing 1- 5 of 5

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat