Uvex™ by Sperian Stealth® Safety Goggles
Sleek style, comfort and protection all in one
  • Low-profile design and indirect venting system for chemical splash and impact protection Scratch-resistant and ideal for most indoor work HydroShield long-life antifog coating helps keep lenses clear Frame and lens are impact resistant
The soft elastomer construction of these goggles conforms to the face for a secure, comfortable and gap-free fit. Its elastomer seal stays soft, pliable and comfortable for the life of the frame. The goggles also feature a headband made of fabric or neoprene, a synthetic polymer that’s resistant to oil, heat and weathering.
There also is an Over-the-Glass (OTG) version of the goggle designed to fit comfortably over prescription eyewear. The safety goggles are clear and made of polycarbonate. They provide greater than 99.9% protection from harmful UV rays and come in other lens tints, too.
Compliance: ANSI Z87.1-2015 and CSA Z94.3 standards.
$20.60 - $24.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Frame Color Lens Type Description
Availability Pricing
Uvex by Honeywell S39610HS Stealth Safety Goggles, Teal Body, Neoprene Headband, Clear, HydroShield Antifog Lens; 1/EA EW-86497-93 Mfr # S39610HS
COMPARE
Teal Standard Safety Goggles, Teal Body, Neoprene Headband, Clear, HydroShield Antifog Lens; 1/EA
$20.60USD / Each
Uvex by Honeywell S3970HS Stealth Safety Goggles, Over-the-Glass, Navy Body, Neoprene Headband, Clear, HydroShield Antifog Lens; 1/EA EW-86497-94 Mfr # S3970HS
COMPARE
Navy Over-the-Glass Safety Goggles, Over-the-Glass, Navy Body, Neoprene Headband, Clear, HydroShield Antifog Lens; 1/EA
$24.50USD / Each