United Electric NEMA 4X Temperature Switches
FEATURES
  • Legendary UE reliability
  • Wideselection of temperature ranges
  • Internal reference scales and adjustment lock
  • Enclosure design permits safe and easy access for wiring
  • Choice of local or remote mount
  • NEMA 4X die cast aluminum enclosure, epoxy powder coated, gasketed, captive screws
$406.25 - $530.00USD / Each
5 variations of this product are available.
See All Product Options Below
5 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
United Electric B100-120 Temperature Switch 0/225F Local Mount, Nema-4x EW-39008-15 Mfr # B100-120
COMPARE
225 0 Temperature Switch 0/225F Local Mount, Nema-4x
$440.00USD / Each
United Electric B100-121 Temperature Switch 200/425F Local Mount, Nema-4x EW-39008-16 Mfr # B100-121
COMPARE
425 200 Temperature Switch 200/425F Local Mount, Nema-4x
$406.25USD / Each
United Electric E100-2BSB Nema 4x Temperature Switch, 30/250F; Remote Mount EW-39008-18 Mfr # E100-2BSB
COMPARE
250 30 Nema 4x Temperature Switch, 30/250F; Remote Mount
$487.50USD / Each
United Electric E100-3BS Nema 4x Temperature Switch, 100/400F; Remote Mount EW-39008-19 Mfr # E100-3BS
COMPARE
400 100 Nema 4x Temperature Switch, 100/400F; Remote Mount
$530.00USD / Each
United Electric E100-8BC Nema 4x Temperature Switch, 350/640F; Remote Mount EW-39008-22 Mfr # E100-8BC
COMPARE
640 350 Nema 4x Temperature Switch, 350/640F; Remote Mount
$431.50USD / Each