Cole-Parmer® Ultrasonic Processor Cup Horn for 500- and 750-Watt Ultrasonic Processors
Process a sample in isolation from the probe
  • Cooling water can be circulated through the cup during operation
Use this cup horn to process a sample in absolute isolation from the probe. The horn is especially useful for processing samples that must be kept in a sealed vessel, such as pathogens. The ultrasonic vibrations are transmitted through the water in the cup causing cavitation in the sample vessel.
$96.00 - $2,160.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Tip Diameter (cm) Tip Diameter (in) Description
Availability Pricing
COMPARE
6.35 2.5 Ultrasonic Processor Cup Horn, 2-1/2\" Dia
$2,160.00USD / Each
COMPARE
0.47752 0.188 Ultrasonic Processor Cup Horn Microtube Holder
$96.00USD / Each