Ultimate Acid Eater
Eliminates the need for powders, sorbent pads, booms, and socks, as well as caustics
Ultimate Acid Eater neutralizes hazardous acids in a quick, safe, and effective manner by transforming them into disposable organic salts. This material will also neutralize dangerous acid vapors that pose safety concerns. It has a 1:1 neutralization ratio or less with a variety of acids. pH color indicator additive enables you to monitor the neutralization process without pH instruments. Neutralizer is easy to apply on vertical surfaces, hard-to-reach areas, and permeable surfaces. Safe to use and store, this nonhazardous, noncorrosive, biodegradable, and environmentally safe product, dries quickly and will not leave any residues or stains. It also eliminates disposal issues and cost as well as safety hazards associated with acid spills.
$288.25 - $447.25USD / Case of 2
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Clift Industries 2001-025 Ultimate Acid Eater; 2.5 Gal. Bottle; 2 Bottles/Case EW-86295-01 Mfr # 2001-025
COMPARE
Ultimate Acid Eater; 2.5 Gal.; 2 Packs/case
$447.25USD / Case of 2
Clift Industries 2002-005 Ultimate Acid Eater Spill Kit, 2 Gallon; 1/Cs EW-86295-02 Mfr # 2002-005
COMPARE
Ultimate Acid Eater Spill Kit, 2 Gallon; 1/Cs
$288.25USD / Each