Cole-Parmer® Two-Stage Pressure Regulators
Ideal for your noncorrosive analytical applications
  • Won’t contaminate high-purity gases
  • Two-stage regulators (Cv = 0.044) compensate for variations in supply pressure
  • Only 0.06 psi outlet change per 100 psi inlet change
Regulators feature 316 stainless steel diaphragms. O-rings of PTFE ensure leak-free operation. Bar stock brass bodies minimize dead space. Automatic reseating valve extends regulator service life. Dual sintered filters reduce gas velocity when opening high pressure cylinders. Chrome plated finish prevents tarnish.
ITEM(S) ON SALE
$613.70USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
COMPARE
Two-Stage Pressure Regulator, 250 SCFH Capacity, 350 CGA Fitting
$613.70USD / Each
$722.00USD / Each
COMPARE
Two-Stage Pressure Regulator, 250 SCFH Capacity, 580 CGA Fitting
$613.70USD / Each
$722.00USD / Each
COMPARE
Two-Stage Pressure Regulator, 250 SCFH Capacity, 590 CGA Fitting
$613.70USD / Each
$722.00USD / Each
COMPARE
Two-Stage Pressure Regulator, 450 SCFH Capacity, 580 CGA Fitting
$613.70USD / Each
$722.00USD / Each
COMPARE
Two-Stage Pressure Regulator, 450 SCFH Capacity, 590 CGA Fitting
$613.70USD / Each
$722.00USD / Each
COMPARE
Two-Stage Pressure Regulator, 900 SCFH Capacity, 350 CGA Fitting
$613.70USD / Each
$722.00USD / Each
COMPARE
Two-Stage Pressure Regulator, 900 SCFH Capacity, 580 CGA Fitting
$613.70USD / Each
$722.00USD / Each
COMPARE
Two-Stage Pressure Regulator, 900 SCFH Capacity, 590 CGA Fitting
$613.70USD / Each
$722.00USD / Each
Showing 1- 8 of 8