Two-Stage General-Purpose Regulators
Two-stage regulators reduce cylinder pressure in two stages. Use first stage to approach working pressure; use second stage to permit precise adjustment or working pressure. Second stage is overload protected.
ITEM(S) ON SALE
$593.30 - $612.00USD / Each
10 variations of this product are available.
See All Product Options Below
10 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (SCFH) Max Delivery Pressure (psig) Max Inlet Pressure (PSI)
Availability Pricing
COMPARE
600 50 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
600 50 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
600 50 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
600 50 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
600 50 3000
$612.00USD / Each
$720.00USD / Each
COMPARE
1200 100 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
1200 100 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
1200 100 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
1200 100 3000
$608.60USD / Each
$716.00USD / Each
COMPARE
1200 100 3000
$593.30USD / Each
$698.00USD / Each
Showing 1- 10 of 10