Turbidity Standard Solutions Kits
Ensure accurate readings by calibrating your turbidimeter
  • Designed especially for your Cole-Parmer®, Hach, LaMotte®, or HF turbidimeter
Kits include a 250-mL bottle of primary standard, a secondary standard-filled cuvette for each of the NTU values listed, and two empty glass cuvettes. All cuvettes are designed to fit your brand-name meter.
$518.50 - $901.50USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
Compareitem specifications
NTU Value
Availability Pricing
COMPARE
0.5, 5.0, 40
$654.50USD / Each
GFS Chemicals Turbidity Standards Kit for Hach 2100 N/AN Meters; 0/20/200/1000/4000 NTU EW-08391-24 Mfr # 85551
COMPARE
0, 20, 200, 1000, 4000
$901.50USD / Each
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro 100 Meters, 0, 10, 1000 NTU EW-08391-29 Mfr # 85555
COMPARE
0, 10, 1000
$561.50USD / Each
GFS Chemicals Turbidity Standards Kit for HF Scientific Micro TPW meter, 0, 10, 1000 NTU EW-08391-70 Mfr # 86226
COMPARE
0, 10, 1000
$518.50USD / Each