TSI IAQ-Calc Indoor Air Quality Meters
Don’t let drift affect readings
  • Low-drift NDIR CO2 sensor for stable, accurate readings
  • Ergonomic, over-molded case design
  • Sampling function records multiple point measurements
Model 7515 is a cost-effective meter for carbon dioxide (CO2) measurements. Models 7525 and 7545 simultaneously measure and data log multiple parameters. Model 7525 measures CO2, temperature, humidity, and calculates dew point, wet bulb temperature and % outside air. Model 7545 adds carbon monoxide (CO).
$788.50 - $2,899.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
TSI 7515 Carbon Dioxide Meter, 0 to 5000 ppm EW-86384-05 Mfr # 7515
COMPARE
Carbon Dioxide Meter, 0 to 5000 ppm
$788.50USD / Each
TSI 7545 Advanced IAQ Meter; Datalogging, CO and CO2, with Humidity EW-86384-15 Mfr # 7545
COMPARE
Advanced IAQ Meter; Datalogging, CO and CO2, with Humidity
$2,899.00USD / Each
TSI 7525 CO2/Temperature/Humidty, with data logging EW-86384-16 Mfr # 7525
COMPARE
CO2/Temperature/Humidty, with data logging
$2,620.00USD / Each