TSI® Alnor® Electronic Balancing Tools
Multiple measurement tools in one package
  • Detachable—can also function as a stand-alone meter
  • Auto-zeroing pressure sensor saves time
  • Automatic density correction increases reading accuracy
  • Up to 5 simultaneous measurements on large, backlit graphic display
  • Intuitive menu structure for easy setup and operation
  • Bluetooth® bidirectional communication for transferring data or remote polling
The Alnor® EBT731 Balometer® capture hood and Alnor EBT730 micromanometer are modular air balancing tools engineered to meet your most demanding HVAC testing, adjusting, balancing, and commissioning requirements — perfect for setting up and testing fume hoods, too!
$2,228.00 - $3,940.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Air Velocity Resolution Max Air Velocity (ft/min) Description
Availability Pricing
TSI EBT730 () Digital Micromanometer EW-05949-49 Mfr # EBT730
COMPARE
1 ft/min (0.01 m/s) 15000 TSI () Digital Micromanometer
$2,228.00USD / Each
TSI EBT731 Electronic Balancing Capture Hood EW-05949-51 Mfr # EBT731
COMPARE
1 ft/min (0.01 m/s) 15000 Electronic Balancing Capture Hood
$3,940.00USD / Each