Troemner Class-F Avoirdupois Stainless Steel Hook on Top Test Weights
Stainless Steel Hook Weights are used in a variety of applications such as pressure, torque, and tensile strength testing
Troemner’s Stainless Steel Hook weights are manufactured from 300 series stainless steel. Each weight has the nominal value stamped into the top side of the weight.
Hook Top Only - Hook Top Weights are flat on top with a stationary hook in the center of the weight
$127.00 - $451.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Troemner 12006-S 10 lb Class F Stainless Steel Hook Top Weight with No Cert EW-01034-01 Mfr # 12006-S
COMPARE
10 lb, Class F Stainless Steel Hook Top Weight with No Certificate
$395.00USD / Each
Troemner 12010-S 2 lb Class F Stainless Steel Hook Top Weight with No Cert EW-01034-02 Mfr # 12010-S
COMPARE
2 lb, Class F Stainless Steel Hook Top Weight with No Certificate
$170.00USD / Each
Troemner 12012-S 1 lb Class F Stainless Steel Hook Top Weight with No Cert EW-01034-03 Mfr # 12012-S
COMPARE
1 lb, Class F Stainless Steel Hook Top Weight with No Certificate
$127.00USD / Each
Troemner 12006-ST 10 lb, Class F Stainless Steel Hook Top Weight with Traceable Certificate EW-01034-14 Mfr # 12006-ST
COMPARE
10 lb, Class F Stainless Steel Hook Top Weight with Tracable Certificate
$451.00USD / Each
Troemner 12010-ST 2 lb, Class F Stainless Steel Hook Top Weight with Traceable Certificate EW-01034-15 Mfr # 12010-ST
COMPARE
2 lb, Class F Stainless Steel Hook Top Weight with Tracable Certificate
$240.00USD / Each
Troemner 12012-ST 1 lb, Class F Stainless Steel Hook Top Weight with Traceable Certificate EW-01034-16 Mfr # 12012-ST
COMPARE
1 lb, Class F Stainless Steel Hook Top Weight with Tracable Certificate
$191.00USD / Each
Troemner 12014-ST 0 0.5 lb, Class F Stainless Steel Hook Top Weight with Traceable Certificate EW-01034-17 Mfr # 12014-ST
COMPARE
0.5 lb, Class F Stainless Steel Hook Top Weight with Tracable Certificate
$180.00USD / Each
Showing 1- 7 of 7