Troemner Class-7 Economical Cylindrical Balance Weights
ASTM Class 7 - For rough basic weighing operations
  • Two-piece weights with adjusting cavities
Economical Weights are economical, quality weights. Individual weights are available from 1 mg through 2000 g. Weights are adjusted to tolerances specified by ANSI/ASTM E617 Class 7.
Economical Class 7 weights are manufactured from stainless steel and are cylindrical in shape for easy gripping. Weights 1 g to 100 g are of one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight. Weights 200 g through 2 kg have an adjusting cavity on the top that is sealed with a plug. Economical Stainless Steel Weights 1 g and above are perfectly flat on top to stack easily on center. Weights 500 mg and lower are made of stainless steel or aluminum and are flat, one-piece construction with one side turned up to make them easy to handle with forceps. 500 mg and 200 mg weights are made of stainless steel while weights 100 mg and below are made of aluminum.
Suggested Markets for Economical Stainless Steel Weights include, but are not limited to, educational markets
$19.80 - $243.00USD / Each
23 variations of this product are available.
See All Product Options Below
23 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g) Description
Availability Pricing
Troemner CLASS 7 1g Class 7 Economical S/S Cyl Indrical Weight EW-01028-42 Mfr # 61013S
COMPARE
1 1g Class 7 Economical S/S Cyl Indrical Weight
$19.80USD / Each
Troemner CLASS 7 2g Class 7 Economical S/S Cyl Indrical Weight EW-01028-44 Mfr # 61023S
COMPARE
2 2g Class 7 Economical S/S Cyl Indrical Weight
$20.30USD / Each
Troemner CLASS 7 5g Class 7 Economical S/S Cyl Indrical Weight EW-01028-46 Mfr # 61053S
COMPARE
5 5g Class 7 Economical S/S Cyl Indrical Weight
$20.30USD / Each
Troemner CLASS 7 10g Class 7 Economical S/S Cyl Indrical Weight EW-01028-48 Mfr # 61014S
COMPARE
10 10g Class 7 Economical S/S Cyl Indrical Weight
$20.50USD / Each
Troemner CLASS 7 20g Class 7 Economical S/S Cyl Indrical Weight EW-01028-52 Mfr # 61024S
COMPARE
20 20g Class 7 Economical S/S Cyl Indrical Weight
$19.80USD / Each
Troemner CLASS 7 50g Class 7 Economical S/S Cyl Indrical Weight EW-01028-54 Mfr # 61054S
COMPARE
50 50g Class 7 Economical S/S Cyl Indrical Weight
$24.00USD / Each
Troemner CLASS 7 100g Class 7 Economical S/S Cyl Indrical Weight EW-01028-56 Mfr # 61015S
COMPARE
100 100g Class 7 Economical S/S Cyl Indrical Weight
$24.00USD / Each
Troemner CLASS 7 200g Class 7 Economical S/S Cyl Indrical Weight EW-01028-58 Mfr # 61025S
COMPARE
200 200g Class 7 Economical S/S Cyl Indrical Weight
$33.50USD / Each
Troemner CLASS 7 500g Class 7 Economical S/S Cyl Indrical Weight EW-01028-62 Mfr # 61055S
COMPARE
500 500g Class 7 Economical S/S Cyl Indrical Weight
$55.50USD / Each
Troemner CLASS 7 1000g Class 7 Economical S/S Cyl Indrical Weight EW-01028-63 Mfr # 61016S
COMPARE
1000 1000g Class 7 Economical S/S Cyl Indrical Weight
$96.50USD / Each
Troemner CLASS 7 2000g Class 7 Economical S/S Cyl Indrical Weight EW-01028-64 Mfr # 61026S
COMPARE
2000 2000g Class 7 Economical S/S Cyl Indrical Weight
$170.00USD / Each
Troemner CLASS 7 1g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-75 Mfr # 61013ST
COMPARE
NIST 1 1g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$45.30USD / Each
Troemner CLASS 7 2g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-76 Mfr # 61023ST
COMPARE
NIST 2 2g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$45.30USD / Each
Troemner CLASS 7 5g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-77 Mfr # 30391027
COMPARE
NIST 5 5g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$45.30USD / Each
Troemner CLASS 7 10g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-78 Mfr # 61014ST
COMPARE
NIST 10 10g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$45.30USD / Each
Troemner CLASS 7 20g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-80 Mfr # 61024ST
COMPARE
NIST 20 20g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$45.30USD / Each
Troemner CLASS 7 50g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-81 Mfr # 61054ST
COMPARE
NIST 50 50g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$50.00USD / Each
Troemner CLASS 7 100g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-82 Mfr # 61015ST
COMPARE
NIST 100 100g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$49.00USD / Each
Troemner CLASS 7 200g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-83 Mfr # 61025ST
COMPARE
NIST 200 200g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$54.30USD / Each
Troemner CLASS 7 500g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate EW-01028-84 Mfr # 61055ST
COMPARE
NIST 500 500g Class 7 Economical S/S Cyl Indrical Weight with Traceable Certificate
$94.50USD / Each
Showing 1 - 20 of 23