Troemner Class-2 Analytical Calibration Weights
ASTM Class 2 - Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01 g to 0.001 g.
  • Surfaces are polished to a perfect, mirror like finish
  • Masses are less vulnerable to wear and scratches
  • Low magnetic susceptibility, uniform consistency and high resistance to corrosion
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity and turned up end for easy handling
  • 1g to 5kg weights are two-piece construction with adjusting cavity and handling knob
Appropriate for calibrating high-precision top loading balances with a readability as low as 0.01 g to 0.001 g.
The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.
Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.
Suggested Markets for Analytical Precision Weights include, but are not limited to, Pharmaceutical QC, R&D, Analytical Testing, Chemical Manufacturing, Utilities, Pharmaceutical Bulk Weighing, General Manufacturing.
$128.00 - $4,096.00USD / Each
41 variations of this product are available.
See All Product Options Below
41 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Mass Certification Mass Size (g) Mass Size (mg)
Availability Pricing
Troemner CLASS 2 5kg Analytical Class 2 Weight with NVLAP Certificate EW-01024-56 Mfr # 7010-2W
COMPARE
NVLAP 5000 5000000
$1,892.00USD / Each
Troemner CLASS 2 2kg Analytical Class 2 Weight with NVLAP Certificate EW-01024-61 Mfr # 7012-2W
COMPARE
NVLAP 2000 2000000
$1,244.00USD / Each
Troemner CLASS 2 1kg Analytical Class 2 Weight with NVLAP Certificate EW-01024-67 Mfr # 7013-2W
COMPARE
NVLAP 1000 1000000
$842.00USD / Each
Troemner CLASS 2 500g Analytical Class 2 Weight with NVLAP Certificate EW-01024-73 Mfr # 7014-2W
COMPARE
NVLAP 500 500000
$439.00USD / Each
Troemner CLASS 2 200g Analytical Class 2 Weight with NVLAP Certificate EW-01024-85 Mfr # 7016-2W
COMPARE
NVLAP 200 200000
$333.00USD / Each
Troemner CLASS 2 100g Analytical Class 2 Weight with NVLAP Certificate EW-01025-02 Mfr # 7017-2W
COMPARE
NVLAP 100 100000
$206.00USD / Each
Troemner CLASS 2 50g Analytical Class 2 Weight with NVLAP Certificate EW-01025-16 Mfr # 7018-2W
COMPARE
NVLAP 50 50000
$199.00USD / Each
Troemner CLASS 2 20g Analytical Class 2 Weight with NVLAP Certificate EW-01025-34 Mfr # 7020-2W
COMPARE
NVLAP 20 20000
$188.00USD / Each
Troemner CLASS 2 10g Analytical Class 2 Weight with NVLAP Certificate EW-01025-54 Mfr # 7021-2W
COMPARE
NVLAP 10 10000
$192.00USD / Each
Troemner CLASS 2 5g Analytical Class 2 Weight with NVLAP Certificate EW-01025-69 Mfr # 7022-2W
COMPARE
NVLAP 5 5000
$181.00USD / Each
Troemner CLASS 2 2g Analytical Class 2 Weight with NVLAP Certificate EW-01025-78 Mfr # 7024-2W
COMPARE
NVLAP 2 2000
$172.00USD / Each
Troemner CLASS 2 1g Analytical Class 2 Weight with NVLAP Certificate EW-01025-82 Mfr # 7025-2W
COMPARE
NVLAP 1 1000
$164.00USD / Each
Troemner CLASS 2 500 mg, Analytical Class 2 Weight with NVLAP Certificate EW-01025-87 Mfr # 7026-2W
COMPARE
NVLAP 0.5 500
$147.00USD / Each
Troemner CLASS 2 100 mg, Analytical Class 2 Weight with NVLAP Certificate EW-01025-97 Mfr # 7029-2W
COMPARE
NVLAP 0.1 100
$152.00USD / Each
Troemner CLASS 2 50 mg, Analytical Class 2 Weight with NVLAP Certificate EW-01026-01 Mfr # 7030-2W
COMPARE
NVLAP 0.05 50
$147.00USD / Each
Troemner CLASS 2 20 mg, Analytical Class 2 Weight with NVLAP Certificate EW-01026-05 Mfr # 7032-2W
COMPARE
NVLAP 0.02 20
$147.00USD / Each
Troemner CLASS 2 10 mg, Analytical Class 2 Weight with NVLAP Certificate EW-01026-09 Mfr # 7033-2W
COMPARE
NVLAP 0.01 10
$147.00USD / Each
Troemner CLASS 2 5 mg, Analytical Class 2 Weight with NVLAP Certificate EW-01026-24 Mfr # 7034-2W
COMPARE
NVLAP 0.005 5
$147.00USD / Each
Troemner CLASS 2 1 mg, Analytical Class 2 Weight with NVLAP Certificate EW-01026-48 Mfr # 7037-2W
COMPARE
NVLAP 0.001 1
$147.00USD / Each
Troemner 7010-2T 5 kg Class 2 Analytical Weight with Traceable Cert EW-01031-53 Mfr # 7010-2T
COMPARE
NIST 5000 5000000
$1,735.00USD / Each
Showing 1 - 20 of 41