Troemner Class-1 Analytical Calibration Weight Sets
ASTM Class 1 - Can be used as a reference standard in calibrating other weights and is appropriate for calibrating high-precision analytical balances with a readability as low as0. 1 mg to 0.01 mg .
  • Masses are less vulnerable to wear and scratches
  • Low magnetic susceptibility, uniform consistency and high resistance to corrosion
  • 1mg to 500mg weights are one-piece construction with no adjusting cavity and turned up end for easy handling
  • 1g to 5kg weights are two-piece construction with adjusting cavity and handling knob
Appropriate for calibrating high-precision analytical balances with readability as low as 0.1 mg to 0.01 mg.
The weights are manufactured of the finest stainless steels available, including Troemner Alloy 8 Stainless Steel, which is available exclusively through Troemner. This unique material possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion. Alloy 8 is the finest grade of stainless steel available and is similar to material originally specified in the manufacture of 1 kg state lab primary standards.
Weights are designed so the specifications on surface finish,markings, construction, material density, and magnetic properties become more restrictive as the weight tolerances
decrease in order to enable the weight to maintain its tolerance over time. These weights are designed for your most demanding weighing applications.
Suggested Markets for Analytical Precision Weights include, but are not limited to,Manufacturing QC, Secondary Calibration Labs, State Laboratories, Pharmaceutical QC, R&D, Analytical Testing, Chemical Manufacturing, Utilities.
$1,267.00 - $6,418.00USD / Each
28 variations of this product are available.
See All Product Options Below
28 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Contents of Set Mass Certification Mass Class
Availability Pricing
Troemner 7226-1 100g to 10 mg, Analytical Class 1 Weight Set EW-01022-85 Mfr # 7226-1
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg; polycarbonate case None Class 1
$1,371.00USD / Each
Troemner 7233-1W 50g-1 mg Class 1 Mass Set, NVLAP Accredited Certificate EW-01023-04 Mfr # 7233-1W
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NVLAP Class 1
$2,803.00USD / Each
Troemner 7228-1W 100g-1 mg Class 1 Mass Set, NVLAP Accredited Certificate EW-01023-18 Mfr # 7228-1W
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; polycarbonate case NVLAP Class 1
$2,996.00USD / Each
Troemner 7217-1W 1kg-1 mg Class 1 Mass Set, NVLAP Accredited Certificate EW-01023-27 Mfr # 7217-1W
COMPARE
1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg; wood case and polycarbonate case NVLAP Class 1
$5,728.00USD / Each
Troemner 7214-1T 2 kg-1 g Class 1 Analytical Set with Traceable Cert EW-01032-51 Mfr # 7214-1T
COMPARE
2 kg, 1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST Class 1
$5,043.00USD / Each
Troemner 7215-1T 2 kg-1 mg Class 1 Analytical Set with Traceable Cert EW-01032-52 Mfr # 7215-1T
COMPARE
2 kg, 1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NIST Class 1
$5,781.00USD / Each
Troemner 7216-1T 1 kg-1 g Class 1 Analytical Set with Traceable Cert EW-01032-53 Mfr # 7216-1T
COMPARE
1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST Class 1
$4,013.00USD / Each
Troemner 7217-1T 1 kg-1 mg Class 1 Analytical Set with Traceable Cert EW-01032-54 Mfr # 7217-1T
COMPARE
1 kg, 500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NIST Class 1
$5,074.00USD / Each
Troemner 7218-1T 500 g-1 g Class 1 Analytical Set with Traceable Cert EW-01032-55 Mfr # 7218-1T
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST Class 1
$2,917.00USD / Each
Troemner 7219-1T 500 g-1 mg Class 1 Analytical Set with Traceable Cert EW-01032-56 Mfr # 7219-1T
COMPARE
500 g, (2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NIST Class 1
$4,035.00USD / Each
Troemner 7220-1T (2) 200 g-1 g Class 1 Analytical Set w/ Traceable Cert EW-01032-57 Mfr # 7220-1T
COMPARE
(2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST Class 1
$1,772.00USD / Each
Troemner 7221-1T (2) 200 g-1 mg Class 1 Analytical Set w/ Traceable Cert EW-01032-58 Mfr # 7221-1T
COMPARE
(2) 200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NIST Class 1
$2,944.00USD / Each
Troemner 7222-1T 200 g-1 g Class 1 Analytical Set with Traceable Cert EW-01032-59 Mfr # 7222-1T
COMPARE
200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST Class 1
$1,543.00USD / Each
Troemner 7223-1T 200 g-1 mg Class 1 Analytical Set with Traceable Cert EW-01032-60 Mfr # 7223-1T
COMPARE
200 g, 100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NIST Class 1
$2,550.00USD / Each
Troemner 7224-1T 100 g-1 g Class 1 Analytical Set with Traceable Cert EW-01032-61 Mfr # 7224-1T
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST Class 1
$1,267.00USD / Each
Troemner 7226-1T 100 g-10 mg Class 1 Analytical Set with Traceable Cert EW-01032-62 Mfr # 7226-1T
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg NIST Class 1
$1,981.00USD / Each
Troemner 7228-1T 100 g-1 mg Class 1 Analytical Set with Traceable Cert EW-01032-63 Mfr # 7228-1T
COMPARE
100 g, 50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NIST Class 1
$2,264.00USD / Each
Troemner 7229-1T 50 g-1 g Class 1 Analytical Set with Traceable Cert EW-01032-64 Mfr # 7229-1T
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g NIST Class 1
$1,269.00USD / Each
Troemner 7231-1T 50 g-10 mg Class 1 Analytical Set with Traceable Cert EW-01032-65 Mfr # 7231-1T
COMPARE
50 g, (2) 20 g, 10 g, 5 g, (2) 2 g, 1 g, 500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg NIST Class 1
$1,798.00USD / Each
Troemner 7240-1T 500 mg-1 mg Class 1 Analytical Set with Traceable Cert EW-01032-67 Mfr # 7240-1T
COMPARE
500 mg, (2) 200 mg, 100 mg, 50 mg, (2) 20 mg, 10 mg, 5 mg, (2) 2 mg, 1 mg NVLAP Class 1
$1,268.00USD / Each
Showing 1 - 20 of 28