This product is now discontinued, View Related Products
Transmitters With DIN Connector/Compression Fitting
Made of 17-4 PH stainless steel with all-welded SS back end
  • Select transducers with a 3-ft (1-m) cable or a DIN connector/compression fitting
Transducers offer exceptional long-term stability and enhanced accuracy. Typical applications include load weighing systems, compressor and dispensing equipment, lube oil pressure equipment, and factory automated equipment. RFI/EMI and ESD protection circuits exceed CE requirements protecting against noise, voltage spikes and static discharge. State-of-the-art ASIC chip in each transducer provides greater linearity correction than traditional thermal compensation methods.
Call For Price
16 variations of this product are available.
See All Product Options Below
16 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
COMPARE
±0.15% full-scale
Call For Price
COMPARE
±0.15% full-scale 7.5
Call For Price
COMPARE
±0.15% full-scale 15
Call For Price
COMPARE
±0.15% full-scale 30
Call For Price
COMPARE
±0.15% full-scale 60
Call For Price
COMPARE
±0.15% full-scale 100
Call For Price
COMPARE
±0.15% full-scale 150
Call For Price
COMPARE
±0.15% full-scale 200
Call For Price
COMPARE
±0.15% full-scale 300
Call For Price
COMPARE
±0.15% full-scale 500
Call For Price
COMPARE
±0.15% full-scale 1000
Call For Price
COMPARE
±0.15% full-scale 1500
Call For Price
COMPARE
±0.15% full-scale 2000
Call For Price
COMPARE
±0.15% full-scale 3000
Call For Price
COMPARE
±0.15% full-scale 5000
Call For Price
COMPARE
±0.15% full-scale 6000
Call For Price
Showing 1- 16 of 16