Traceable® T-Bar Waterproof Food Thermometers with Calibration
T-bar shape design make it easy to pierce through frozen and semisolid foods
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory│Waterproof (IP67 rating)—ideal for food and beverage applications and in washdown areas│Rugged, shockproof and drop tested to withstand a fall of 5 feet (1.5 meters)│°F/°C switchable
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$32.50 - $68.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
536 -58 Calibrated T-Bar Waterproof Food Thermometer; ±1°C accuracy (-20 to 100°C)
$32.50USD / Each
COMPARE
536 -58 Calibrated T-Bar Waterproof Food Thermometer Ultra; ±0.4°C Accuracy at Tested Points
$68.00USD / Each