Traceable® Precision Sentry™ Digital Thermometers
Simultaneously display Min/Max and current temperature
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory NIST-traceable calibration
  • Simple—one-key—Min/Max resets
  • Use either the external or internal temperature sensor
  • Toggle between °F/°C
  • View battery status at a glance
These digital thermometers constantly monitor and display current, minimum, and maximum temperatures. Resetting the minimum and maximum temperature recorded is a simple one-button operation.
Choose between three external probe types; bullet, bottle, or vaccine-bottle to suit your application needs. Probes feature a 9-ft (2.7-m) cable for placement in hoods, refrigerators/freezers, incubators, or submerged in a bath. Unplugging the external sensor will automatically activate the internal/ambient sensor. Ambient sensor has a range of -4 to 122°F (-20 to 50°C) with the same accuracy.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$63.50 - $82.50USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° C) Max Temperature (° F) Description
Availability Pricing
COMPARE
60 140 Calibrated Digital Thermometer, Bullet Probe
$63.50USD / Each
COMPARE
60 140 Calibrated Digital Thermometer, Bottle Probe
$82.50USD / Each
COMPARE
60 140 Calibrated Digital Thermometer, Vaccine-Bottle Probe
$82.50USD / Each