Traceable® Lollipop™ Water-Resistant Thermometers with Calibration
Shockproof thermometers update reading every second
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Stainless-steel probe is resistant to most laboratory chemicals
  • One-key operation recalls MIN/MAX temperature readings
  • °F/°C switchable
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$36.00 - $79.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Description
Availability Pricing
COMPARE
572 -58 Calibrated Water-Resistant Thermometer; ±1.0°C Accuracy (-20 to 100°C)
$36.00USD / Each
COMPARE
572 -58 Calibrated Waterproof Thermometer Ultra; ±0.4°C Accuracy at Tested Points
$79.00USD / Each