Traceable® High-Accuracy Refrigerator/Freezer Thermometers with Calibration
Triple-display—view MIN, MAX, and current temperature simultaneously
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory│Resolution of 0.01° and accuracy of ±0.6°F (±0.3°C)│Visual and audible alarms signal continuously until reset
Easily and accurately monitor temperature in most scientific, industrial, or general-purpose applications. Built-in alarms alert continuously until reset. Unit displays the exact time and date when thermometer alarms are triggered. Program the alarm in 0.1° increments. Meters that accept two probes are ideal for monitoring two unique areas that are near each other. Probe comes with a microcable that permits a door to close on it.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$106.00 - $165.00USD / Each
7 variations of this product are available.
See All Product Options Below
7 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Number Of Probe Channels Description
Availability Pricing
COMPARE
2 Calibrated High-Accuracy Fridge/Freezer Thermometer; 2 Bullet Probes
$106.00USD / Each
COMPARE
2 Calibrated High-Accuracy Fridge/Freezer Thermometer; 2 Bottle Probes
$131.00USD / Each
COMPARE
1 Calibrated High-Accuracy Fridge/Freezer Thermometer; 1 Stainless Steel Probe
$132.00USD / Each
COMPARE
2 Calibrated High-Accuracy Fridge/Freezer Thermometer; 2 Stainless Steel Probes
$165.00USD / Each
COMPARE
1 Calibrated High-Accuracy Fridge/Freezer Thermometer; 1 Bottle Probe
$118.00USD / Each
COMPARE
1 Calibrated High-Accuracy Fridge/Freezer Thermometer; 1 Vaccine Bottle Probe
$119.00USD / Each
COMPARE
2 Calibrated High-Accuracy Fridge/Freezer Thermometer; 2 Vaccine Bottle Probes
$135.00USD / Each
Showing 1- 7 of 7