Traceable® Extreme-Accuracy Digital Thermometer with Calibration
Perfect for monitoring critical samples, quality control checks, as well as routine measurements
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
  • Resolution is 0.01°
  • Displays date and time of min/max temperature
  • High and low settable alarms│Unit conforms to ITS-90, International Temperature Standard
Four Traceable International Standards Digital thermometers provide the exact readings required for critical requirements, quality control checks, and routine measurements. Units read 0.00, 25.00, 37.00, or a combination of all three with a resolution of 0.01° and an accuracy ±0.05°C within ±2°C of standard. Units read in °C and °F.
Impervious stainless-steel probe
Stainless-steel probe supplied with units shows the slightest temperature change. Probe is triple purpose for liquid, air/gas, and semisolids. Accurate readings with tip penetration of 1/3-inch. Probe has a diameter of 1/8-inch, stem length of 6-1/4 inches, overall length of 9 inches and a cable length of 5 feet. Absolute high-precision thermometer is ideal for measuring temperatures in water baths, cuvettes, gas systems, reagents, standards solutions, ovens, incubators, pharmaceuticals, petroleum products, foods, and wastewater. The display shows minimum/maximum and current temperatures. MIN/MAX monitors readings overnight, on weekends, or for any time period and displays the exact time and date when the MIN/MAX temperature occurred. Alarm provides two visual (LEDs) and two audio alerts when temperature rises above or falls below high and low set points. Unit displays the exact time and date when dual thermometer alarms are triggered. Alarms are programmable in 0.1° increments. Visual and audible alarms signal continuously (for days) even if temperature returns to non-alarm range. First thermometer ever designed to permit user one-button calibration.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$134.00 - $142.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Meter Measurement Type Number Of Probe Channels
Availability Pricing
COMPARE
70 Digital Indicator 1
$134.00USD / Each
COMPARE
70 Digital Indicator 1
$134.00USD / Each
COMPARE
70 Digital Indicator 1
$134.00USD / Each
COMPARE
70 Digital Indicator 1
$142.00USD / Each