Traceable® Excursion-Trac™ Data Logging Refrigerator/Freezer Thermometers with Calibration
Rolling memory retains most recent 525,600 temperature observations
  • Includes NIST-traceable certificate from our A2LA-accredited (1750.01) calibration laboratory
  • Memory can be cleared after downloading data and alarm events
  • Set limits for and store up to 10 unique alarm events
  • Smart-Alarm™ with visual/audible alarm continues to alert even if conditions return to normal range
  • Triple-display—shows high, low, and current temperature simultaneously
  • Can upload a USB downloaded file and store on a TraceableLIVE® cloud account
Easily monitor temperatures overnight or for any time period—minimum and maximum temperatures are stored until the memory is cleared. Set high and low alarms in 1° increments; alarm sounds when temperature rises above or falls below set points. The alarm tracking feature stores data for up to 10 different alarm events detailing when the alarm state occurred, as well as when the unit returned within range. Alarm state indicator includes visual LEDs, audio alerts, and flashing LCD segments.
Transfer data recorded in a CSV format to any computer via a USB flash drive—no software is required. Simply clear memory after downloading data to USB drive. Unit continues to record new data while downloading old data. Other status indicators include low-battery, memory full, USB data transfer, and active alarm state. All probes are attached to a 10-ft (3-m) microcable that permits a door to close on it.
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$169.00 - $470.00USD / Each
15 variations of this product are available.
See All Product Options Below
15 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° F) Number Of Probe Channels Description
Availability Pricing
COMPARE
-58 1 Calibrated Data Logging Thermometer; 1 Bottle Probe
$178.00USD / Each
COMPARE
-58 2 Calibrated Data Logging Thermometer; 2 Bottle Probes
$185.00USD / Each
COMPARE
-58 1 Calibrated Data Logging Thermometer; 1 Bullet Probe
$169.00USD / Each
COMPARE
-58 2 Calibrated Data Logging Thermometer; 2 Bullet Probes
$184.00USD / Each
COMPARE
-58 1 Calibrated Data Logging Thermometer; 1 Stainless Steel Probe
$192.00USD / Each
COMPARE
-58 2 Calibrated Data Logging Thermometer; 2 Stainless Steel Probes
$206.00USD / Each
COMPARE
-58 2 Calibrated Data Logging Thermometer; 1 Bottle/1 Bullet Probe
$190.00USD / Each
COMPARE
-58 1 Calibrated Datalogging Thermometer; 1 Vaccine Bottle Probe
$179.00USD / Each
COMPARE
-58 2 Calibrated Datalogging Thermometer; 2 Vaccine Bottle Probes
$192.00USD / Each
COMPARE
-58 1 Calibrated Datalogging Thermometer; 1 Plastic Bottle Probe
$171.00USD / Each
COMPARE
-58 2 Calibrated Datalogging Thermometer; 2 Plastic Bottle Probes
$185.00USD / Each
COMPARE
-130 1 Calibrated Datalogging Low-Temp Thermometer; 1 Stainless Steel Probe
$245.00USD / Each
COMPARE
-130 2 Calibrated Datalogging Low-Temp Thermometer; 2 Stainless Steel Probes
$264.00USD / Each
COMPARE
-328 1 Calibrated Datalogging Cryogenic Thermometer; 1 Stainless Steel Probe
$412.00USD / Each
COMPARE
-328 2 Calibrated Datalogging Cryogenic Thermometer; 2 Stainless Steel Probes
$470.00USD / Each
Showing 1- 15 of 15