Traceable® Cryogenic Wi-Fi Data Loggers Compatible with TraceableLIVE® Cloud Service
Ensure viability of irreplaceable samples from your mobile devices
  • Connect to your data via smartphone, tablet, or PC using TraceableLIVE wireless capability*
  • View current Liquid Nitrogen (LN2) conditions anywhere—cloud-based interface sends email, text, and push notifications
  • No software required—web, Android™, Apple® iOS data visibility
  • Includes NIST-Traceable certificate from our A2LA accredited (1750.01) calibration laboratory
TraceableLIVE Wireless Technology allows multiple users to stay connected and monitor critical environments 24/7 wherever you go.
Monitoring priceless biological samples has never been easier! This liquid nitrogen (LN2) data logging thermometer with Wi-Fi connectivity monitors cryogenic storage temperature and streams data continuously to mobile devices via the cloud-based interface. For those in the lab, audible and visual alarms alert when conditions are outside of the safe range.
Once setup is complete you can securely monitor parameters 24/7, control your alarms remotely, view your data logging history, run reports in real time, and give access to other team members. No local software is required to use the cloud-based data interface. Get mobile push notifications, emails, or texts for out of range conditions, connectivity interruptions, low-battery condition or if your device is due for calibration. Large tripe-line LCD displays current and Min/Max readings.
Each data logger is calibrated according to NIST at four points (-196°C, -80°C, 0°C, 100°C) ensuring accuracy and reliability of measurements. Never lose sensitive samples to liquid nitrogen leaks or other catastrophic accidents again!
Use the included hook-and-loop strip to mount the probe. Mount the logger with the included magnetic strip or use the built-in bench stand to place on countertop. A low-cost subscription is required for TraceableLIVE® use.
Plus the NIST-traceable precalibration means you get reliable measurements right out of the box, saving you time and money!
Traceable® Products have an individually-numbered Traceable® Certificate provided with each unit, that assures accuracy from our ISO/IEC 17025:2017(1750.01) calibration laboratory accredited by A2LA. It indicates traceability of measurements to the SI units through NIST or other recognized national measurement institutes (NMI) that are signatories to the CIPM Mutual Recognition Agreement. Each product goes through an intense vetting process before it can carry the Traceable name, thus saving you both time and money by not having to calibrate separately.
$477.00 - $533.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Min Temperature (° C) Min Temperature (° F) Number of Channels
Availability Pricing
COMPARE
-200 -328 1
$477.00USD / Each
COMPARE
-200 -328 2
$533.00USD / Each