Torbal FSA Torgue Cap Tester
1000Hz Data Capture
  • Wireless data transmitter
  • 2.75" LCD display with backlight
  • USB, RS232, and analog l/O interface
  • Removable MicroSD card
  • Onscreen Stats and analytics
  • Onscreen graphing and histogram
Cap tester offers superior performance for the most demanding production or quality control applications. Engineered with high output loadcells the FSA TORQUE delivers accurate and repeatable torque measurements. With six modes and applications you can test any container or jar ranging from 0.79" (20 mm) to 7.09" (180 mm) in diameter. Addjustable steel posts with a non-slip rubber sleeve assures a firm grip on a wide range of container shapes and sizes. A die cast metal housing provides a stable platform for the torgue gauge and overall durability for a long life in demanding environments.
$1,655.00 - $1,765.00USD / Each
3 variations of this product are available.
See All Product Options Below
3 PRODUCT OPTIONS
Compareitem specifications
Capacity (lb-in) Capacity (N-m) Sampling Rate
Availability Pricing
Torbal FSA2 Digital Cap Torque Tester, 17 lb in capacity EW-25304-73 Mfr # FSA2
COMPARE
17 2 1000Hz
$1,655.00USD / Each
Torbal FSA5 Digital Cap Torque Tester, 44 lb in capacity EW-25304-74 Mfr # FSA5
COMPARE
44 5 1000Hz
$1,710.00USD / Each
Torbal FSA10 Digital Cap Torque Tester, 100 lb in capacity EW-25304-75 Mfr # FSA10
COMPARE
100 10 1000Hz
$1,765.00USD / Each