Tintometer Comparator 3000 Benchtop Color Grading Systems
Economical, visual color measurement with three-field determination
  • Available with the most popular color scales
  • Quick comparison of sample to glass color standards
Convenient benchtop comparator is a three-field instrument for visually determining the color of samples by direct comparison with colored glass filters. With a 3-section field of view, the sample and two consecutive glasses on the color scale are viewed simultaneously, making it easier to achieve the optimal color match. For rapid color grading within predetermined color limits, the glasses can be set to the two limiting colors, making it easy to see if the sample is within tolerance.
$5,840.00USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Description
Availability Pricing
Lovibond AF228 Gardner Benchtop Gardner Comparator EW-99562-38 Mfr # 345010
COMPARE
Benchtop Gardner Comparator
$5,840.00USD / Each