Cole-Parmer® Essentials Self-Adhering Cable Zip Tie Mounts
Installs without tools or hardware
  • Secures bundles to surfaces where mounting holes cannot be drilled
  • High-strength adhesive securely sticks to solid, clean surfaces
  • Speeds assembly – cable ties slide easily into mounts from any of four sides
  • Conforms to minor surface irregularities
Nylon cable zip tie mounts have a strong adhesive to help support cable and wire bundles and are perfect for many areas where hardware mounting is not possible. Simply peel off backing and stick to glass, metal, plastic, or other solid, clean surface. They will conform to minor surface irregularities. The four-sided design allows cables to be inserted fast and easily from any side. These adhesive mounts have a UL 94 V-2 flammability rating, an operating temperature of -30 °C (-22 °F) to 75 °C (167 °F), and a minimum installation temperature of 0 °C (32 °F).
$25.00 - $29.50USD / Bag of 100
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Length (in) Compatible With Width (in)
Availability Pricing
COMPARE
0.84 18 to 40 lb cable ties 0.84
$25.00USD / Bag of 100
COMPARE
1.09 40 to 50 lb cable ties 1.09
$29.50USD / Bag of 100