ThermoProbe Precision RTD Digital Handheld Stem Thermometers
Used to replace any precision glass stem or compact digital thermometers
  • Easy-to-read indicator
  • Highly accurate RTD probe
Whether you’re taking a simple temperature measurement, verifying the accuracy of a temperature sensor or performing a complex procedure, these handheld thermometers give you a safe, portable, and accurate solution for all of your temperature applications. Excellent alternative for glass thermometers — with no concerns of mercury and accurate for the entire temperature range. Typical industries include laboratory, petroleum, pharmaceutical, and food.
The probe and digital readout are fixed together, so they are calibrated as a system and are extremely accurate over the full temperature range with selectable resolution of 0.1, 0.01, or 0.001 (depending on model). The stabilization indicator shows you if your temperature is trending up or down and lets you know when your temperature source is stable enough for an accurate measurement. These units also feature Min/Max/Avg reading, selectable temperature scale (°F/°C), low-battery indicator, and auto-off function.
$535.00 - $1,338.53USD / Each
8 variations of this product are available.
See All Product Options Below
8 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° F) Min Temperature (° F) Stem Length (in)
Availability Pricing
ThermoProbe Intrinsically Safe Stick Thermometer; 8" Long Stem EW-90026-09 Mfr # TL3-A-8
COMPARE
400 -40 8
$828.18USD / Each
ThermoProbe Intrinsically Safe Stick Thermometer; 12" Long Stem EW-90026-10 Mfr # TL3-A-12
COMPARE
400 -40 12
$828.18USD / Each
ThermoProbe Intrinsically Safe Stick Thermometer with Horizontal Display; 8" Stem EW-90026-11 Mfr # TL3-W- 8
COMPARE
400 -40 8
$978.76USD / Each
ThermoProbe Intrinsically Safe Stick Thermometer with Horizontal Display; 12" Stem EW-90026-12 Mfr # TL3-W-12
COMPARE
400 -40 12
$978.76USD / Each
ThermoProbe Intrinsically Safe Stick Thermometer; High-Accuracy, 8" Long Stem EW-90026-13 Mfr # TL3-R-8
COMPARE
300 -4 8
$1,338.53USD / Each
ThermoProbe Intrinsically Safe Stick Thermometer; High-Accuracy, 12" Long Stem EW-90026-14 Mfr # TL3-R-12
COMPARE
300 -4 12
$1,338.53USD / Each
ThermoProbe Handheld Stem Thermometer with 8" (203 mm) Long Stem EW-90026-15 Mfr # TL3-8
COMPARE
400 -40 8
$535.00USD / Each
ThermoProbe Handheld Stem Thermometer with 12" (304 mm) Long Stem EW-90026-16 Mfr # TL3-12
COMPARE
400 -40 12
$535.00USD / Each
Showing 1- 8 of 8