Thermo Scientific™ Forma™ Series II Water-Jacketed CO2 Incubators
Durable triple wall cabinet construction ensures optimal temperature uniformity
  • TC sensor for constant temperature and humidity applications
  • IR sensor for changing temperature and humidity applications
These incubators provide security of samples in event of power loss as they hold the temperature longer than air-jacketed models. They feature a temperature-adjustable heated door which keeps the inner glass door free of condensation for improved viewing of cultures. Audible and visual alarms indicate temperature deviations from 0.5° to 5° below or above set point, and
CO2 concentration deviations of 0.5% to 5% below or above set point. Validatable in-chamber HEPA air filtration system minimizes the risk of airborne contaminants entering the incubator upon door openings, so you can maintain your cultures in Class 100 air quaility conditions. You can set a HEPA filter replacement reminder or the default notifies you every 6 months. Built-in RS-485 port can be used to transfer temperature and CO2 data to a printer. Easy-to-read LED display scans between actual temperature and CO2 level.
$11,600.00 - $13,600.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (cu ft) Power (VAC) Description
Availability Pricing
Thermo Scientific Forma 3110 Water Jacketed CO2 Incubator; TC 120 EW-39320-04 Mfr # 3110
COMPARE
6.5 90 to 125 Water Jacketed CO2 Incubator; TC 120
$11,600.00USD / Each
Thermo Scientific Forma 3120 Water Jacketed CO2 Incubator; IR 120 EW-39320-08 Mfr # 3120
COMPARE
6.5 90 to 125 Water Jacketed CO2 Incubator; IR 120
$13,600.00USD / Each