Thermo Scientific Thermolyne Compact Benchtop Muffle Furnaces
Digital control offers simple economical operation
  • Ideal for ignition tests, gravimetric analysis, and heat treating steel parts
  • Feature single set point digital control
  • Digital LED displays chamber temperature
Furnaces feature ceramic fiber insulation for fast heat-up and reduced energy consumption—rise time to 1100°>C takes only 50 minutes. Embedded heating elements on three sides give an improved temperature gradient. Drop-down door serves as a convenient shelf for loading and unloading samples. A safety interlock switch disconnects power to heating elements whenever the door is opened. Chambers are constructed of heavy-duty welded steel. Order optional hearth plate separately.
$1,595.00 - $1,945.00USD / Each
4 variations of this product are available.
See All Product Options Below
4 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Capacity (Liters) Power (VAC)
Availability Pricing
Thermo Scientific Thermolyne FB1315M Compact Benchtop Muffle Furnace; 76 cu in; 120V EW-33900-00 Mfr # FB1315M
COMPARE
1.3 120
$1,595.00USD / Each
Thermo Scientific Thermolyne FB1310M-33 Compact Benchtop Muffle Furnace; 76 cu in; 240V EW-33900-05 Mfr # FB1310M-33
COMPARE
1.3 240
$1,700.00USD / Each
Thermo Scientific Thermolyne FB1415M Compact Benchtop Muffle Furnace; 127.5 cu in; 120V EW-33900-10 Mfr # FB1415M
COMPARE
2.1 120
$1,945.00USD / Each
Thermo Scientific Thermolyne FB1410M-33 Compact Benchtop Muffle Furnace; 127.5 cu in; 240V EW-33900-15 Mfr # FB1410M-33
COMPARE
2.1 240
$1,890.00USD / Each