This product is now discontinued, View Related Products
Thermo Scientific Thermolyne Atmosphere-Controlled Muffle Furnaces
Built-in flowmeter with valve allows easy adjustment of airflow
  • Stainless steel manifold introduces pre-warmed gases
  • Exhaust port allows for easy ventilation—no fume hood needed
These programmable, atmosphere-controlled muffle furnaces are ideal for coal and coke ashing procedures. Easily adjust the flow rate of air (0 to 80 L/min) with the built-in flowmeter located in the front of the furnace.
Stainless steel (SS) manifold pre-warms incoming gases to provide minimum temperature gradient. There is no need to place furnace under a fume hood—connect the stainless steel tubing to exhaust port on top of the furnace for proper ventilation; order tubing kit separately.
The furnace has an eight-segment (four ramp and four dwell) programmable controller and a digital LED that lets you view both the actual and set temperatures. Overtemperature protection cuts off power to the heating elements when chamber temperature exceeds set point.
Call For Price
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Capacity (Liters) Power (VAC)
Availability Pricing
Thermo Scientific Thermolyne F6028C-60-80 Programmable Ashing Furnace with Purge/Vent; 208V EW-33900-40 Mfr # F6028C-60-80
COMPARE
14.2 208
Call For Price
Thermo Scientific Thermolyne F6020C-33-60-80 Programmable Ashing Furnace with Purge/Vent; 240V EW-33900-45 Mfr # F6020C-33-60-80
COMPARE
14.2 240