This product is now discontinued, View Related Products

Thermo Scientific Revco CxF Ultra-Low Temperature Chest Freezers

Reliable sample protection and versatile storage configurations
Revco CxF chest freezers feature superior protection for all your sample storage. Available in two temperature options, -40°C (-40°F) or -86°C (-122.8°F), and in four convenient sizes. Available racking solutions allow you to optimize your storage needs. An easy-to-use microprocessor control panel features a message center with a 12-character LED display showing system status and alarm messages. Access codes help ensure set point security. Optional chart recorder (factory installed) provides continuous visible monitoring.


Heavy-gauge, cold-rolled steel exterior with a powder-coat finish resists chipping and rust and stainless steel interior is durable and simple to clean. The 5" (12.7 cm) thick foamed-in-place polyurethane insulation provides excellent thermal qualities. Interior covers minimize sample exposure to outside conditions while inner sub-lids protect bulk samples from exposure during retrievals. Other features include an easy-to-remove washable air filter, lockable lid handle, and RS-232 and four 20 mA outputs for independent monitoring devices. The locking heavy-duty casters provide smooth mobility.

Call For Price

22 variations of this product are available.

See All Product Options Below
22 PRODUCT OPTIONS
Compareitem specifications
Freezer Capacity (cu ft) Power (Hz) Power (VAC)
Availability Pricing
Thermo Scientific Revco ULT350-10-A CxF Chest Freezer, -40C, 3 cu ft; 115V/60Hz EW-44147-01 Mfr # ULT350-10-A
COMPARE
3 60 115
Call For Price
Thermo Scientific Revco ULT350-10-V CxF Chest Freezer, -40C, 3 cu ft; 230V/50Hz EW-44147-02 Mfr # ULT350-10-V
COMPARE
3 50 230
Call For Price
Thermo Scientific Revco ULT1350-10-A CxF -40°C Chest Freezer, 12.7 cubic feet, 115V/60Hz EW-44147-03 Mfr # ULT1350-10-A
COMPARE
12.7 60 115
Call For Price
Thermo Scientific Revco ULT1350-10-D CxF Chest Freezer, -40C, 12.7 cu ft; 208V/60Hz EW-44147-04 Mfr # ULT1350-10-D
COMPARE
12.7 60 208 / 230
Call For Price
Thermo Scientific Revco ULT1350-10-V CxF -40°C Chest Freezer, 12.7 cubic feet, 230V/50Hz EW-44147-05 Mfr # ULT1350-10-V
COMPARE
12.7 50 230
Call For Price
Thermo Scientific Revco ULT1750-10-A CxF -40°C Chest Freezer, 17 cubic feet, 115V/60Hz EW-44147-06 Mfr # ULT1750-10-A
COMPARE
17 60 115
Call For Price
Thermo Scientific Revco ULT1750-10-D CxF Chest Freezer, -40C, 17cu ft; 208V/60Hz EW-44147-07 Mfr # ULT1750-10-D
COMPARE
17 60 208 / 230
Call For Price
Thermo Scientific Revco ULT1750-10-V CxF -40°C Chest Freezer, 17 cubic feet, 230V/50Hz EW-44147-08 Mfr # ULT1750-10-V
COMPARE
17 50 230
Call For Price
Thermo Scientific Revco ULT2050-10-A CxF -40°C Chest Freezer, 20 cubic feet, 115V/60Hz EW-44147-09 Mfr # ULT2050-10-A
COMPARE
20 60 115
Call For Price
Thermo Scientific Revco ULT2050-10-D CxF Chest Freezer, -40C, 20 cu ft; 208V/60Hz EW-44147-10 Mfr # ULT2050-10-D
COMPARE
20 60 115
Call For Price
Thermo Scientific Revco ULT2050-10-V CxF -40°C Chest Freezer, 20 cubic feet, 230V/50Hz EW-44147-11 Mfr # ULT2050-10-V
COMPARE
20 50 230
Call For Price
Thermo Scientific Revco ULT390-10-A CxF -86°C Chest Freezer, 3 cubic feet, 115V/60Hz EW-44147-12 Mfr # ULT390-10-A
COMPARE
3 60 115
Call For Price
Thermo Scientific Revco ULT390-10-V CxF -86°C Chest Freezer, 3 cubic feet, 230V/50Hz EW-44147-13 Mfr # ULT390-10-V
COMPARE
3 50 230
Call For Price
Thermo Scientific Revco ULT1390-10-A CxF -86°C Chest Freezer, 12.7 cubic feet, 115V/60Hz EW-44147-14 Mfr # ULT1390-10-A
COMPARE
12.7 60 115
Call For Price
Thermo Scientific Revco ULT1390-10-D CxF Chest Freezer, -86C, 12.7 cu ft; 208V/60Hz EW-44147-15 Mfr # ULT1390-10-D
COMPARE
12.7 60 115
Call For Price
Thermo Scientific Revco ULT1390-10-V CxF -86°C Chest Freezer, 12.7 cubic feet, 230V/50Hz EW-44147-16 Mfr # ULT1390-10-V
COMPARE
12.7 50 230
Call For Price
Thermo Scientific Revco ULT1790-10-A CxF -86°C Chest Freezer, 17 cubic feet, 115V/60Hz EW-44147-17 Mfr # ULT1790-10-A
COMPARE
17 60 115
Call For Price
Thermo Scientific Revco ULT1790-10-D CxF Chest Freezer, -86C, 17 cu ft; 208V/60Hz EW-44147-18 Mfr # ULT1790-10-D
COMPARE
17 60 115
Call For Price
Thermo Scientific Revco ULT1790-10-V CxF -86°C Chest Freezer, 17 cubic feet, 230V/50Hz EW-44147-19 Mfr # ULT1790-10-V
COMPARE
17 50 230
Call For Price
Thermo Scientific Revco ULT2090-10-A CxF -86°C Chest Freezer, 20 cubic feet, 115V/60Hz EW-44147-20 Mfr # ULT2090-10-A
COMPARE
20 60 115
Call For Price
Showing 1 - 20 of 22

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat