Thermo Scientific Precision General-Purpose Digital Water Baths
Easily operate and monitor bath with icon-based LCD
  • Over-temperature safety circuitry with audible alarm protects your work
  • Auto-on and auto-off timers allow you to optimize operation schedules
  • Easily clean the chamber with its seamless stainless steel interior
  • Save valuable bench space with compact baths
Thermo Scientific Precision general-purpose baths are rugged, high-performance baths ideal for a wide range of lab applications. Capacities range from 2 to 28 L, including shallow and dual-chamber models. These baths feature an easy-to-read icon-based, graphical display which allows you to save commonly used settings with four temperature presets. Polycarbonate lid provides full view of bath contents and reduces heat loss.
Choose from models with or without thermal beads . The beads replace water to reduce the risk of contamination. Stainless steel covers sold separately.
Note: Thermal beads do not provide the same performance as water; see manual for additional instructions.
$1,215.00 - $6,260.00USD / Each
6 variations of this product are available.
See All Product Options Below
6 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max Temperature (° C) Reservoir Capacity (L) Description
Availability Pricing
Thermo Scientific Precision TSGP02 General Purpose Water Baths, 2L EW-03710-90 Mfr # TSGP02
COMPARE
90 2 General Purpose Water Baths, 2L
$1,215.00USD / Each
Thermo Scientific Precision TSGP2S General Purpose Water Bath, 2L Shallow EW-03710-91 Mfr # TSGP2S
COMPARE
100 2 General Purpose Water Bath, 2L Shallow
$1,215.00USD / Each
Thermo Scientific Precision TSGP05 General Purpose Water Baths, 5L EW-03710-92 Mfr # TSGP05
COMPARE
100 5 General Purpose Water Baths, 5L
$1,765.00USD / Each
Thermo Scientific Precision TSGP28 General Purpose Water Baths, 28L EW-03710-96 Mfr # TSGP28
COMPARE
100 28 General Purpose Water Baths, 28L
$3,435.00USD / Each
Thermo Scientific Precision TSGP15D General Purpose Water Baths, 5/10L Dual EW-03710-97 Mfr # TSGP15D
COMPARE
100 10 General Purpose Water Baths, 5/10L Dual
$2,775.00USD / Each
Thermo Scientific Precision TSGP28PMO5 General Purpose Water Baths, 28L, with 24L Beads EW-03711-02 Mfr # TSGP28PMO5
COMPARE
100 28 General Purpose Water Baths, 28L, with 24L Beads
$6,260.00USD / Each