Thermo Scientific Owl Double-Sided Large-Vertical Electrophoresis Systems
Larger-size cassette gives you the maximum number of samples and separation length
  • Upper buffer chamber is extended, providing quick and easy gel casts
  • Built-in cooling system circulates cold tap water for fast run times and flat, even banding patterns
  • Simple clamping mechanism for trouble-free loading of glass cassettes and precast gels
$1,860.00 - $3,255.00USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Buffer Solution Required (mL) Glass Length (cm) Glass Width (cm)
Availability Pricing
Thermo Scientific P9DS-1 Owl Large-Vertical Double-Sided Cell, 16 x 14 cm gel size EW-28554-05 Mfr # P9DS-1
COMPARE
700 14 16
$1,860.00USD / Each
Thermo Scientific P10DS-1 Standard Double-Sided Vertical Cell, 20 x 20 cm gel size EW-28554-15 Mfr # P10DS-1
COMPARE
1400 20 20
$3,255.00USD / Each