Thermo Scientific Orion Star A200 Series pH/mV/RmV/Temperature Benchtop Meter
Durability and dependability make it the ideal choice for every lab, every sample, every day
  • Multilanguage interface supports English, Spanish, French, Italian, German and Chinese
  • Improved stability algorithm provide better measurement accuracy
  • Ensure temperature accuracy with ATC temperature calibration
  • Nonvolatile memory holds up to 2000 data points with time date stamp
  • Individual method passwords provide better security
  • Option of AC power or four AA batteries (sold separately)
Thermo Scientific Orion Star A211 pH, mV, RmV and temperature meter features an intuitive user interface with plain language screen prompts. A large, backlit graphic LCD, shows measurement and temperature along with electrode status, time/date, sample ID, user ID, and calibration data.
Up to five point automatic calibration with USA/NIST and DIN buffers, auto-buffer recognition buffers, and recalibration feature to fix calibration errors. GLP-related features include 2000 data points with time/date stamp and electrode verification. Meter utilizes RS-232 or USB for connectivity and includes data analysis software.
ITEM(S) ON SALE
$1,089.00 - $1,468.80USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
Compareitem specifications
Max pH (pH) Max Temperature (° C) Description
Availability Pricing
Thermo Scientific STAR A211 Star A211 pH Benchtop Meter Only EW-58825-22 Mfr # STARA2110
COMPARE
20 105 Star A211 pH Benchtop Meter Only
$1,089.00USD / Each
$1,210.00USD / Each
Thermo Scientific STAR A211 Star A211 pH Benchtop Meter Kit
Always in Stock
EW-58825-24 Mfr # STARA2115
COMPARE
20 105 Star A211 pH Benchtop Meter Kit
$1,468.80USD / Each
$1,632.00USD / Each